Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

meso-1,2-Bis(p-tolyl)ethylenediamine, 98%, Thermo Scientific™

Catalog No. AC375271000
Change view
Click to view available options
Quantity:
100 mg
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC375271000 100 mg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC375271000 Supplier Thermo Scientific Chemicals Supplier No. 375271000

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 50764-59-5
Molecular Formula C16H20N2
Molecular Weight (g/mol) 240.35
InChI Key UQUZLWQGLCLOBD-IYBDPMFKSA-P
Synonym 1r,2s-1,2-bis 4-methylphenyl ethane-1,2-bis aminium, 1s,2r-2-azaniumyl-1,2-bis 4-methylphenyl ethyl azanium
PubChem CID 7017343
IUPAC Name [(1R,2S)-2-azaniumyl-1,2-bis(4-methylphenyl)ethyl]azanium
SMILES CC1=CC=C(C=C1)C(C(C2=CC=C(C=C2)C)[NH3+])[NH3+]

Specifications

Melting Point 113°C to 117°C
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula (CH3C6H4CH(NH2)-)2
Quantity 100 mg
Formula Weight 240.35
Percent Purity 98%
Chemical Name or Material meso-1, 2-Bis(p-tolyl)ethylenediamine
Hazard Category GHS Signal Word: Danger

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.