accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3ONNHNBr(E/Z)

CAS RN 50783-82-9

IUPAC Name: 6-[2-(5-bromopyridin-2-yl)hydrazin-1-ylidene]-3-(dimethylamino)cyclohexa-2,4-dien-1-one
Synonyms: 6-[2-(5-bromopyridin-2-yl)hydrazin-1-ylidene]-3-(dimethylamino)cyclohexa-2,4-dien-1-one
Molecular Weight (g/mol): 321.18
Molecular Formula: C13H13BrN4O
InChi Key: GXYVDXVMTIAMFF-UHFFFAOYSA-N
SMILES: CN(C)C1=CC(=O)C(C=C1)=NNC1=NC=C(Br)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results