accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaHOOOOO

CAS RN 50795-24-9

IUPAC Name: sodium 2-(carboxymethoxy)acetate
Synonyms: sodium 2-(carboxymethoxy)acetate
Molecular Weight (g/mol): 156.07
Molecular Formula: C4H5NaO5
InChi Key: WESJBTCFPDPYGH-UHFFFAOYSA-M
SMILES: [Na+].OC(=O)COCC([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results