Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydrogen Diglycolate, MP Biomedicals

Catalog No. ICN218036 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN218036 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN218036 Supplier MP Biomedicals, Inc Supplier No. 0521803610

Chemical Identifiers

CAS 50795-24-9
Molecular Formula C4H5NaO5
Molecular Weight (g/mol) 156.07
InChI Key WESJBTCFPDPYGH-UHFFFAOYSA-M
Synonym sodium hydrogen diglycolate, acetic acid, 2,2'-oxybis-, monosodium salt, diglycolic acid monosodium salt, acetic acid, 2,2'-oxybis-, sodium salt 1:1, monosodium diglycolate, 2-carboxymethoxy acetic acid, sodium salt, sodium hydrogen oxydi acetate, sodium hydrogen oxydiacetate, wesjbtcfpdpygh-uhfffaoysa-m, sodium 2-carboxymethoxy acetate
PubChem CID 23688988

Specifications

Quantity 10 g
Chemical Name or Material Sodium Hydrogen Diglycolate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.