accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOCH3

CAS RN 50802-52-3

IUPAC Name: 4-(hexyloxy)phenyl 4-pentylbenzoate
Synonyms: 4-(hexyloxy)phenyl 4-pentylbenzoate
Molecular Weight (g/mol): 368.52
Molecular Formula: C24H32O3
InChi Key: QOSDDNTYRMLJLY-UHFFFAOYSA-N
SMILES: CCCCCCOC1=CC=C(OC(=O)C2=CC=C(CCCCC)C=C2)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results