Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hexyloxyphenyl 4-Pentylbenzoate 99.0+%, TCI America™
SDP

Catalog No. P08971G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P08971G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P08971G Supplier TCI America Supplier No. P08971G

Chemical Identifiers

CAS 50802-52-3
Molecular Formula C24H32O3
Molecular Weight (g/mol) 368.517
MDL Number MFCD00059437
InChI Key QOSDDNTYRMLJLY-UHFFFAOYSA-N
Synonym 4-Amylbenzoic Acid 4-Hexyloxyphenyl Ester, 4-Pentylbenzoic Acid 4-Hexyloxyphenyl Ester
PubChem CID 3084945
IUPAC Name (4-hexoxyphenyl) 4-pentylbenzoate
SMILES CCCCCCOC1=CC=C(C=C1)OC(=O)C2=CC=C(C=C2)CCCCC

Specifications

Color White
Quantity 1 g
Formula Weight 368.52
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hexyloxyphenyl 4-Pentylbenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.