CAS RN 5089-72-5
IUPAC Name:
(2-aminoethyl)[3-(triethoxysilyl)propyl]amine
Synonyms:
(2-aminoethyl)[3-(triethoxysilyl)propyl]amine
Molecular Weight (g/mol):
264.44
Molecular Formula:
C11H28N2O3Si
InChi Key:
INJVFBCDVXYHGQ-UHFFFAOYSA-N
SMILES:
CCO[Si](CCCNCCN)(OCC)OCC
1
–
1
of
1
results
1
3-(2-Aminoethylamino)propyltriethoxysilane 96.0+%, TCI America™
CAS: 5089-72-5 Molecular Formula: C11H28N2O3Si Molecular Weight (g/mol): 264.44 MDL Number: MFCD00138097 InChI Key: INJVFBCDVXYHGQ-UHFFFAOYSA-N Synonym: N-(3-Triethoxysilylpropyl)ethylenediamine PubChem CID: 78772 IUPAC Name: (2-aminoethyl)[3-(triethoxysilyl)propyl]amine SMILES: CCO[Si](CCCNCCN)(OCC)OCC