Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(2-Aminoethylamino)propyltriethoxysilane 96.0+%, TCI America™
SDP

Catalog No. A192325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A192325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A192325G Supplier TCI America Supplier No. A192325G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5089-72-5
Molecular Formula C11H28N2O3Si
Molecular Weight (g/mol) 264.44
MDL Number MFCD00138097
InChI Key INJVFBCDVXYHGQ-UHFFFAOYSA-N
Synonym N-(3-Triethoxysilylpropyl)ethylenediamine
PubChem CID 78772
IUPAC Name (2-aminoethyl)[3-(triethoxysilyl)propyl]amine
SMILES CCO[Si](CCCNCCN)(OCC)OCC

Specifications

Color Yellow
Boiling Point 156°C
Quantity 25 g
Formula Weight 264.44
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-(2-Aminoethylamino)propyltriethoxysilane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.