accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrON

CAS RN 50916-55-7

IUPAC Name: 3-(2-bromoacetyl)benzonitrile
Synonyms: 3-(2-bromoacetyl)benzonitrile
Molecular Weight (g/mol): 224.06
Molecular Formula: C9H6BrNO
InChi Key: XWCGNFLHRINYCE-UHFFFAOYSA-N
SMILES: BrCC(=O)C1=CC=CC(=C1)C#N
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)