Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(2-Bromoacetyl)benzonitrile, 95%, Thermo Scientific™

Catalog No. SPB07593DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SPB07593DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SPB07593DA Supplier Thermo Scientific Chemicals Supplier No. SPB07593DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 50916-55-7
Molecular Formula C9H6BrNO
Molecular Weight (g/mol) 224.06
MDL Number MFCD00833267
InChI Key XWCGNFLHRINYCE-UHFFFAOYSA-N
Synonym 3-2-bromoacetyl benzonitrile, 3-cyanophenacyl bromide, 3-cyanophenacylbromide, 2-bromo-3'-cyanoacetophenone, 3-2-bromoacetyl benzenecarbonitrile, 3-bromoacetyl benzonitrile, 3-2-bromo-acetyl-benzonitrile, benzonitrile, 3-bromoacetyl, 3-bromoacetyl-benzonitrile
PubChem CID 2735867
IUPAC Name 3-(2-bromoacetyl)benzonitrile
SMILES BrCC(=O)C1=CC=CC(=C1)C#N

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Linear Formula BrCH2C(O)C6H4CN
Quantity 1 g
Formula Weight 224.06
Percent Purity 95%
Chemical Name or Material 3-(2-Bromoacetyl)benzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.