accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2HOOOHO

CAS RN 50930-79-5

IUPAC Name: aniline; benzene-1,2-dicarboxylic acid
Synonyms: aniline; phthalic acid
Molecular Weight (g/mol): 259.26
Molecular Formula: C14H13NO4
InChi Key: QGPZXNSBZMHHSR-UHFFFAOYSA-N
SMILES: NC1=CC=CC=C1.OC(=O)C1=CC=CC=C1C(O)=O
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results