Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aniline Hydrogen Phthalate, MP Biomedicals

Catalog No. ICN201056 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN201056 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN201056 Supplier MP Biomedicals, Inc Supplier No. 0520105610
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 50930-79-5
Molecular Formula C14H13NO4
Molecular Weight (g/mol) 259.261
InChI Key QGPZXNSBZMHHSR-UHFFFAOYSA-N
Synonym aniline hydrogen phthalate, aniline phthalate, aniline; phthalic acid, 1,2-benzenedicarboxylic acid, compd. with benzenamine 1:1, c8h6o4.c6h7n, phthalic acid, compound with aniline 1:1, aniline hydrogenphthalate, aniline hydrogen phtalate, aniline phthalate solution, phthalic acid aniline salt
PubChem CID 170940
IUPAC Name aniline;phthalic acid
SMILES C1=CC=C(C=C1)N.C1=CC=C(C(=C1)C(=O)O)C(=O)O

Specifications

Quantity 10 g
Formula Weight 259.26
Chemical Name or Material Aniline Hydrogen Phthalate
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.