accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CN

CAS RN 50966-65-9

IUPAC Name: 1-octyl-1H-pyrrole
Synonyms: 1-octylpyrrole
Molecular Weight (g/mol): 179.31
Molecular Formula: C12H21N
InChi Key: AXCMRJJCPRARIT-UHFFFAOYSA-N
SMILES: CCCCCCCCN1C=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results