Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-n-Octylpyrrole 95.0+%, TCI America™
SDP

Catalog No. O02425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O02425G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O02425G Supplier TCI America Supplier No. O02425G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 50966-65-9
Molecular Formula C12H21N
Molecular Weight (g/mol) 179.307
MDL Number MFCD00191473
InChI Key AXCMRJJCPRARIT-UHFFFAOYSA-N
PubChem CID 10631204
IUPAC Name 1-octylpyrrole
SMILES CCCCCCCCN1C=CC=C1

Specifications

Color Yellow
Boiling Point 130°C
Quantity 5 g
Formula Weight 179.31
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-n-Octylpyrrole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.