accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaNaH2NSOOOSOOO

CAS RN 50976-35-7

IUPAC Name: disodium 6-aminonaphthalene-1,3-disulfonate
Synonyms: disodium 6-aminonaphthalene-1,3-disulfonate
Molecular Weight (g/mol): 347.27
Molecular Formula: C10H7NNa2O6S2
InChi Key: YKOUFOHKVHDKRS-UHFFFAOYSA-L
SMILES: [Na+].[Na+].NC1=CC=C2C(=C1)C=C(C=C2S([O-])(=O)=O)S([O-])(=O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results