Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Disodium 6-Amino-1,3-naphthalenedisulfonate Hydrate 95.0+%, TCI America™
SDP

Catalog No. A03405G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A03405G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A03405G Supplier TCI America Supplier No. A03405G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 50976-35-7
Molecular Formula C10H7NNa2O6S2
Molecular Weight (g/mol) 347.267
MDL Number MFCD00070485
InChI Key YKOUFOHKVHDKRS-UHFFFAOYSA-L
Synonym Amino J Acid Disodium Salt, 6-Amino-1,3-naphthalenedisulfonic Acid Disodium Salt, 2-Naphthylamine-5,7-disulfonic Acid Disodium Salt
PubChem CID 170944
IUPAC Name disodium;6-aminonaphthalene-1,3-disulfonate
SMILES C1=CC2=C(C=C(C=C2C=C1N)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Quantity 5 g
Formula Weight 347.27
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Disodium 6-Amino-1,3-naphthalenedisulfonate Hydrate
EINECSNumber (4)-0502
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.