accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NHN

CAS RN 50995-95-4

IUPAC Name: 2-propyl-1H-imidazole
Synonyms: 2-propyl-1H-imidazole
Molecular Weight (g/mol): 110.16
Molecular Formula: C6H10N2
InChi Key: MKBBSFGKFMQPPC-UHFFFAOYSA-N
SMILES: CCCC1=NC=CN1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results