Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Propylimidazole 95.0+%, TCI America™
SDP

Catalog No. P09475G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P09475G 5 g
P094725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P09475G Supplier TCI America Supplier No. P09475G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 50995-95-4
Molecular Formula C6H10N2
Molecular Weight (g/mol) 110.16
MDL Number MFCD00059158
InChI Key MKBBSFGKFMQPPC-UHFFFAOYSA-N
PubChem CID 162617
IUPAC Name 2-propyl-1H-imidazole
SMILES CCCC1=NC=CN1

Specifications

Melting Point 60°C
Color White-Yellow
Boiling Point 152°C
Quantity 5 g
Formula Weight 110.16
Percent Purity ≥95.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Propylimidazole
EINECSNumber (5)-0382
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.