accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOOOH3C

CAS RN 51-03-6

IUPAC Name: 5-{[2-(2-butoxyethoxy)ethoxy]methyl}-6-propyl-2H-1,3-benzodioxole
Synonyms: piperonyl butoxide caswell no. 670 piperonyl butoxide preparation piperonyl butoxide, technical butocide technical piperonyl butoxide pyrenone 606 piperonyl butoxyde piperonyl butoxide (substance) ccris 522
Molecular Weight (g/mol): 338.44
Molecular Formula: C19H30O5
InChi Key: FIPWRIJSWJWJAI-UHFFFAOYSA-N
SMILES: CCCCOCCOCCOCC1=CC2=C(OCO2)C=C1CCC
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (4)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (6)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (1)
  • (2)
  • (4)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (6)
  • (4)
  • (4)
  • (1)