Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Piperonyl Butoxide, 80%, MP Biomedicals

Catalog No. ICN217595 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN217595 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN217595 Supplier MP Biomedicals, Inc Supplier No. 0521759580

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 51-03-6
Molecular Formula C19H30O5
Molecular Weight (g/mol) 338.44
MDL Number MFCD00005842
InChI Key FIPWRIJSWJWJAI-UHFFFAOYSA-N
Synonym piperonyl butoxide, butacide, butocide, ethanol butoxide, pyrenone 606, piperonylbutoxide, 6-propylpiperonyl butyl diethylene glycol ether, butyl carbitol 6-propylpiperonyl ether, 5-2-2-butoxyethoxy ethoxy methyl-6-propylbenzo d 1,3 dioxole, butoxide synergist
PubChem CID 5794
ChEBI CHEBI:32687
IUPAC Name 5-{[2-(2-butoxyethoxy)ethoxy]methyl}-6-propyl-2H-1,3-benzodioxole
SMILES CCCCOCCOCCOCC1=CC2=C(OCO2)C=C1CCC

Specifications

Quantity 100 g
Formula Weight 338.43
Chemical Name or Material Piperonyl Butoxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.