accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClNH2HO

CAS RN 51-19-4

IUPAC Name: hydrogen 2-aminophenol chloride
Synonyms: hydrogen 2-aminophenol chloride
Molecular Weight (g/mol): 145.59
Molecular Formula: C6H8ClNO
InChi Key: QUDZAFKCASPJLX-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NC1=CC=CC=C1O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results