Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Aminophenol Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. A03915G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A03915G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A03915G Supplier TCI America Supplier No. A03915G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 51-19-4
Molecular Formula C6H8ClNO
Quantity 5 g
MDL Number MFCD00035471
InChI Key QUDZAFKCASPJLX-UHFFFAOYSA-N
SMILES [H+].[Cl-].NC1=CC=CC=C1O
IUPAC Name hydrogen 2-aminophenol chloride
Molecular Weight (g/mol) 145.59
PubChem CID 5800
Formula Weight 145.59
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2-Aminophenol Hydrochloride
Show More Show Less
RTECSNumber SJ6069000
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.