accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaOHOIOOII

CAS RN 51-24-1

IUPAC Name: sodium 2-[4-(4-hydroxy-3-iodophenoxy)-3,5-diiodophenyl]acetate
Synonyms: tiratricol
Molecular Weight (g/mol): 643.92
Molecular Formula: C14H8I3NaO4
InChi Key: BBBAIWCYEJXUMH-UHFFFAOYSA-M
SMILES: [Na+].OC1=CC=C(OC2=C(I)C=C(CC([O-])=O)C=C2I)C=C1I
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results