accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 51-41-2

NH2(R)OHOHHO

CAS RN 51-41-2

IUPAC Name: 4-[(1R)-2-amino-1-hydroxyethyl]benzene-1,2-diol
Synonyms: norepinephrine (+)-isomer norepinephrine noradrenaline bitartrate norepirenamine norepinephrine, (+)- nor adrenalin levophed levarterenol renaudin, noradrénaline tartrate noradrenaline acid tartrate
Molecular Weight (g/mol): 169.18
Molecular Formula: C8H11NO3
InChi Key: SFLSHLFXELFNJZ-QMMMGPOBSA-N
SMILES: NC[C@H](O)C1=CC=C(O)C(O)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results