Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Noradrenaline, 98%

Catalog No. AAL0808703
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0808703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0808703 Supplier Thermo Scientific Chemicals Supplier No. L0808703
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 51-41-2 | C8H11NO3 | 169.18 g/mol

L-Noradrenaline serves as an adrenergic neurotransmitter. It acts as demethylated precursor of epinephrine (adrenaline). It is also used to treat people having low blood pressure, as a sympathomimetic drug to treat people in vasodilatory shock states such as septic shock and neurogenic shock. Further, it is used in the preparation of1,2,3,4-tetrahydro-4,6,7-isoquinolinetriol hydrochloride. In addition to this, it is used in the treatment of hypotension, depression, schizophrenia and attention hyperactivity disorders.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Noradrenaline serves as an adrenergic neurotransmitter. It acts as demethylated precursor of epinephrine (adrenaline). It is also used to treat people having low blood pressure, as a sympathomimetic drug to treat people in vasodil
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51-41-2
Molecular Formula C8H11NO3
Molecular Weight (g/mol) 169.18
MDL Number MFCD00025592
InChI Key SFLSHLFXELFNJZ-QMMMGPOBSA-N
Synonym norepinephrine, l-noradrenaline, levarterenol, arterenol, noradrenaline, levophed, --norepinephrine, l-norepinephrine, r-noradrenaline, r-norepinephrine
PubChem CID 439260
ChEBI CHEBI:18357
IUPAC Name 4-[(1R)-2-amino-1-hydroxyethyl]benzene-1,2-diol
SMILES NC[C@H](O)C1=CC=C(O)C(O)=C1

Specifications

Melting Point ∽220°C (decomposition)
Quantity 1 g
UN Number UN2811
Beilstein 4231961
Sensitivity Air and light sensitive
Merck Index 14,6695
Solubility Information Soluble in alkali and dilute hydrochloric acid. Slightly soluble in water,ethanol and diethyl ether.
Optical Rotation −45° (c=5 in 0.5N HCl)
Formula Weight 169.18
Percent Purity 98%
Chemical Name or Material L-Noradrenaline
Show More Show Less
Hazard Category H300
Hazard Statement GHS H Statement
H300
Fatal if swallowed.
Precautionary Statement P264b-P270-P301+P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 200-096-6
RTECSNumber DN5950000
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.