accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNH(R)OHOHHOOH(R)(R)OHHOOOHO

CAS RN 51-42-3

IUPAC Name: (2R,3R)-2,3-dihydroxybutanedioic acid; 4-[(1R)-1-hydroxy-2-(methylamino)ethyl]benzene-1,2-diol
Synonyms: L(+)-tartaric acid; epinephrine
Molecular Weight (g/mol): 333.29
Molecular Formula: C13H19NO9
InChi Key: YLXIPWWIOISBDD-NDAAPVSOSA-N
SMILES: O[C@H]([C@@H](O)C(O)=O)C(O)=O.CNC[C@H](O)C1=CC=C(O)C(O)=C1
Physical Form 
  • (3)
  • (4)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (4)
  • (3)
Percent Purity 
  • (3)
  • (4)
  • (1)
Grade 
  • (1)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (4)
  • (1)
  • (1)
  • (2)
  • (4)
  • (3)
  • (1)
  • (4)
  • (3)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (3)
  • (3)
  • (4)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)