Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ L-(–)-Epinephrine-(+)-bitartrate

Catalog No. 324900100MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-490-0100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 32-490-0100MG Supplier MilliporeSigma™ Supplier No. 324900100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

α,β-Adrenergic receptor agonist.

α,β-Adrenergic receptor agonist. Has positive chronotropic and inotropic effects on heart muscle. Activates adenylate cyclase to generate cAMP from ATP.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 95% by TLC
Target Proteins α,β-Adrenergic receptor agonist
CAS 51-42-3
Molecular Formula C9H13NO3·C4H6O6
Formula Weight 333.3g/mol
Solubility EtOH or H2O
Content And Storage Hygroscopic
Form Solid
Quantity 100 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.