accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 51-45-6

IUPAC Name: 2-(1H-imidazol-5-yl)ethan-1-amine
Synonyms: 1H-imidazole-5-ethanamine
Molecular Weight (g/mol): 111.15
Molecular Formula: C5H9N3
InChi Key: NTYJJOPFIAHURM-UHFFFAOYSA-N
SMILES: NCCC1=CN=CN1
Quantity 
  • (3)
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (1)
  • (3)
Physical Form 
  • (3)
  • (2)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (5)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (3)
  • (3)
  • (2)
  • (3)
  • (1)