Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histamine

Catalog No. AAJ6172706
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6172706 5 g
AAJ6172703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6172706 Supplier Thermo Scientific Chemicals Supplier No. J6172706
Add to Cart

CAS: 51-45-6 | C5H9N3 | 111.15 g/mol

Histamine inhibits the synthesis of IL-2 and γ-IFN in peripheral blood mononuclear cells and lipopolysaccharide-induced synthesis of TNF-α in monocytes via H2 receptor activation. It is a powerful stimulant of gastric secretion, a constrictor of bronchial smooth muscle, a vasodilator, and also a centrally acting neurotransmitter.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Histamine inhibits the synthesis of IL-2 and γ-IFN in peripheral blood mononuclear cells and lipopolysaccharide-induced synthesis of TNF-α in monocytes via H2 receptor activation. It is a powerful stimulant of gastric secretion, a constrictor of bronchial smooth muscle, a vasodilator, and also a centrally acting neurotransmitter.

Solubility
Soluble in water, alcohol, and hot chloroform.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C Hygroscopic. Handle and store under inert gas. Air sensitive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51-45-6
Molecular Formula C5H9N3
Molecular Weight (g/mol) 111.15
MDL Number MFCD00005210,MFCD00128939
InChI Key NTYJJOPFIAHURM-UHFFFAOYSA-N
Synonym histamine, 1h-imidazole-4-ethanamine, 2-4-imidazolyl ethylamine, ergotidine, 2-1h-imidazol-5-yl ethanamine, ergamine, eramin, 5-imidazoleethylamine, theramine, free histamine
PubChem CID 774
ChEBI CHEBI:18295
SMILES NCCC1=CN=CN1

Specifications

Color White
Quantity 5 g
UN Number UN2811
Beilstein 2012
Merck Index 14,4719
Solubility Information Soluble in water,alcohol,and hot chloroform.
IUPAC Name 2-(1H-imidazol-5-yl)ethan-1-amine
Formula Weight 111.15
Physical Form Powder
Chemical Name or Material Histamine
Hazard Category H301-H315-H317-H319-H334-H335
Hazard Statement GHS H Statement
H301-H334-H315-H319-H317-H335
Toxic if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P272-P280-P285-P301+P310-P302+P352-P304+P340-P305+P351+P338-P330-P333+P313-P342+P311-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 200-100-6
RTECSNumber MS1050000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.