accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2N(R)IOIHOIIHOO

CAS RN 51-49-0

IUPAC Name: (2R)-2-amino-3-[4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl]propanoic acid
Synonyms: dextrothyroxine D-T4 thyroid hormone D thyroxine dextrothyroxinum sodium dextrothyroxine
Molecular Weight (g/mol): 776.87
Molecular Formula: C15H11I4NO4
InChi Key: XUIIKFGFIJCVMT-GFCCVEGCSA-N
SMILES: N[C@H](CC1=CC(I)=C(OC2=CC(I)=C(O)C(I)=C2)C(I)=C1)C(O)=O
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)