Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Thyroxine 97%, Thermo Scientific™

Catalog No. AC138950010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC138950010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC138950010 Supplier Thermo Scientific Chemicals Supplier No. 138950010

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51-49-0
Molecular Formula C15H11I4NO4
Molecular Weight (g/mol) 776.87
MDL Number MFCD00063056
InChI Key XUIIKFGFIJCVMT-GFCCVEGCSA-N
Synonym d-thyroxine, dextrothyroxine, choloxin, r-2-amino-3-4-4-hydroxy-3,5-diiodophenoxy-3,5-diiodophenyl propanoic acid, dextrothyroxinum, unii-4w9k63fion, dextrothyroxine sodium, sodium dextrothyroxine, d-t4, chembl559
PubChem CID 8730
ChEBI CHEBI:30659
IUPAC Name (2R)-2-amino-3-[4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl]propanoic acid
SMILES N[C@H](CC1=CC(I)=C(OC2=CC(I)=C(O)C(I)=C2)C(I)=C1)C(O)=O

Specifications

Color Beige to Gray
Loss on Drying 5% max. (105°C)
Infrared Spectrum Authentic
Quantity 1 g
Beilstein 14, IV, 2373
Merck Index 15, 9570
Specific Rotation -17° to -19° (20°C, 589nm) (c=2, EtOH/1N HCl : 4/1)
Formula Weight 776.86
Percent Purity ≥96.0% (on dry substance)
Physical Form Powder
Chemical Name or Material D-Thyroxine, 97%
Show More Show Less
Hazard Category LIGHT SENSITIVE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.