accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 51065-64-6

IUPAC Name: 6-methylhept-2-yne
Synonyms: 6-methylhept-2-yne
Molecular Weight (g/mol): 110.2
Molecular Formula: C8H14
InChi Key: HIEALULIKYDRQN-UHFFFAOYSA-N
SMILES: CC#CCCC(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results