Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Methyl-2-heptyne, 98%, Thermo Scientific™

Catalog No. AAB2088106
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2088106 5 g
AAB2088103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2088106 Supplier Thermo Scientific Chemicals Supplier No. B2088106

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51065-64-6
Molecular Formula C8H14
Molecular Weight (g/mol) 110.20
MDL Number MFCD00041619
InChI Key HIEALULIKYDRQN-UHFFFAOYSA-N
Synonym 6-methyl-2-heptyne, 2-heptyne, 6-methyl, acmc-1asem, hiealulikydrqn-uhfffaoysa, ch3c.equiv.c ch2 2ch ch3 2
PubChem CID 142809
IUPAC Name 6-methylhept-2-yne
SMILES CC#CCCC(C)C

Specifications

Density 0.76
Refractive Index 1.425
Quantity 5 g
UN Number UN3295
Formula Weight 110.2
Percent Purity 98%
Chemical Name or Material 6-Methyl-2-heptyne
Hazard Category H225-H304
Hazard Statement GHS H Statement
H225-H304
Highly flammable liquid and vapour.
May be fatal if swallowed and enters airways.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P301+P310-P303+P361+P353-P331-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.