accessibility menu, dialog, popup

UserName

Viewing profile as user:
N

CAS RN 51069-11-5

IUPAC Name: 4-phenyl-1-azabicyclo[2.2.2]octane
Synonyms: 4-phenylquinuclidine
Molecular Weight (g/mol): 187.29
Molecular Formula: C13H17N
InChi Key: CSKNWIPXDBBWRW-UHFFFAOYSA-N
SMILES: C1CC2(CCN1CC2)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results