Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Phenylquinuclidine, 98%, Thermo Scientific™

Catalog No. AAL15523MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL15523MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL15523MD Supplier Thermo Scientific Chemicals Supplier No. L15523MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Phenylquinuclidine is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51069-11-5
Molecular Formula C13H17N
Molecular Weight (g/mol) 187.286
MDL Number MFCD00797340
InChI Key CSKNWIPXDBBWRW-UHFFFAOYSA-N
PubChem CID 142810
IUPAC Name 4-phenyl-1-azabicyclo[2.2.2]octane
SMILES C1CN2CCC1(CC2)C3=CC=CC=C3

Specifications

Melting Point 123°C to 124°C
Quantity 250 mg
Synonym 4-phenylquinuclidine, 1-azabicyclo 2.2.2 octane, 4-phenyl, 4-phenyl-1-azabicyclo 2.2.2 octane, 4-phenylquinuclidine #, acmc-1akg7, 1-azabicyclo 2.2.2 octane,4-phenyl, 4-phenyl-1-aza-bicyclo 2.2.2 octane, 4-phenyl-1-azabicyclo-2.2.2 octane, 4-phenyl-1-azabicyclo 2.2.2-octane
Solubility Information Insoluble in water.
Formula Weight 187.29
Percent Purity 98%
Chemical Name or Material 4-Phenylquinuclidine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.