accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOHSOOO

CAS RN 51071-75-1

IUPAC Name: 2,4,6-triphenyl-1λ⁴-pyran-1-ylium hydrogen sulfate
Synonyms: 2,4,6-triphenyl-1λ⁴-pyran-1-ylium hydrogen sulfate
Molecular Weight (g/mol): 406.45
Molecular Formula: C23H18O5S
InChi Key: IPEOZISWOGHJJY-UHFFFAOYSA-M
SMILES: OS([O-])(=O)=O.C1=CC=C(C=C1)C1=CC(=[O+]C(=C1)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results