Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Triphenylpyrylium Hydrogensulfate 97.0+%, TCI America™
SDP

Catalog No. T14451G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T14451G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T14451G Supplier TCI America Supplier No. T14451G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 51071-75-1
Molecular Formula C23H18O5S
Molecular Weight (g/mol) 406.45
MDL Number MFCD00012002
InChI Key IPEOZISWOGHJJY-UHFFFAOYSA-M
PubChem CID 11729198
IUPAC Name 2,4,6-triphenyl-1λ⁴-pyran-1-ylium hydrogen sulfate
SMILES OS([O-])(=O)=O.C1=CC=C(C=C1)C1=CC(=[O+]C(=C1)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 271°C
Quantity 1 g
UN Number 1759
Formula Weight 406.45
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,6-Triphenylpyrylium Hydrogensulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.