accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 51077-16-8

CAS RN 51077-16-8

IUPAC Name: 3-[(tert-butoxy)carbonyl]-1,3-thiazolidine-4-carboxylic acid
Synonyms: 3-(tert-butoxycarbonyl)-1,3-thiazolidine-4-carboxylic acid
Molecular Weight (g/mol): 233.28
Molecular Formula: C9H15NO4S
InChi Key: FJWNZTPXVSWUKF-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)N1CSCC1C(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results