accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHCH2

CAS RN 51100-54-0

IUPAC Name: dec-1-en-3-ol
Synonyms: 3-decenol
Molecular Weight (g/mol): 156.27
Molecular Formula: C10H20O
InChi Key: NSFWLHKFTGZFBP-UHFFFAOYNA-N
SMILES: CCCCCCCC(O)C=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results