Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Decen-3-ol 97.0+%, TCI America™
SDP

Catalog No. D326825ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D326825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D326825ML Supplier TCI America Supplier No. D326825ML

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Chemical Identifiers

CAS 51100-54-0
Molecular Formula C10H20O
Molecular Weight (g/mol) 156.269
InChI Key NSFWLHKFTGZFBP-UHFFFAOYSA-N
Synonym 3-Hydroxy-1-decene
PubChem CID 170977
IUPAC Name dec-1-en-3-ol
SMILES CCCCCCCC(C=C)O

Specifications

Color Colorless
Boiling Point 217°C
Quantity 25 mL
Formula Weight 156.27
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Decen-3-ol
EINECSNumber (2)-0258
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.