accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SH2CSH3C

CAS RN 51102-74-0

IUPAC Name: 1,1-bis(methylsulfanyl)ethene
Synonyms: 1,1-bis(methylsulfanyl)ethene
Molecular Weight (g/mol): 120.23
Molecular Formula: C4H8S2
InChi Key: GGCGAESAURTGKY-UHFFFAOYSA-N
SMILES: CSC(=C)SC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results