accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 51128-24-6

IUPAC Name: 4-butoxyphenyl 4-pentylbenzoate
Synonyms: 4-butoxyphenyl 4-pentylbenzoate
Molecular Weight (g/mol): 340.46
Molecular Formula: C22H28O3
InChi Key: LEJYVZMFIRFBCQ-UHFFFAOYSA-N
SMILES: CCCCCC1=CC=C(C=C1)C(=O)OC1=CC=C(OCCCC)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results