Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Butoxyphenyl 4-Pentylbenzoate 99.0+%, TCI America™
SDP

Catalog No. P08961G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P08961G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P08961G Supplier TCI America Supplier No. P08961G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 51128-24-6
Molecular Formula C22H28O3
Molecular Weight (g/mol) 340.463
MDL Number MFCD00059435
InChI Key LEJYVZMFIRFBCQ-UHFFFAOYSA-N
Synonym 4-Amylbenzoic Acid 4-Butoxyphenyl Ester, 4-Pentylbenzoic Acid 4-Butoxyphenyl Ester
PubChem CID 44630445
IUPAC Name (4-butoxyphenyl) 4-pentylbenzoate
SMILES CCCCCC1=CC=C(C=C1)C(=O)OC2=CC=C(C=C2)OCCCC

Specifications

Color White
Quantity 1 g
Formula Weight 340.46
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Butoxyphenyl 4-Pentylbenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.