accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 51149-08-7

IUPAC Name: 3,6-dichloropyridazine-4-carboxylic acid
Synonyms: 3,6-dichloropyridazine-4-carboxylic acid
Molecular Weight (g/mol): 192.98
Molecular Formula: C5H2Cl2N2O2
InChi Key: FRCXPDWDMAYSCE-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(Cl)=NN=C1Cl
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)