CAS RN 51203-49-7
IUPAC Name:
1-titanaspiro[heptacyclo[3.3.0.0¹,³.0¹,⁴.0¹,⁶.0¹,⁷.0²,⁸]octane-1,1'-pentacyclo[2.2.0.0¹,³.0¹,⁵.0²,⁶]hexane]
Synonyms:
1-titanaspiro[heptacyclo[3.3.0.0¹,³.0¹,⁴.0¹,⁶.0¹,⁷.0²,⁸]octane-1,1'-pentacyclo[2.2.0.0¹,³.0¹,⁵.0²,⁶]hexane]
Molecular Weight (g/mol):
204.1
Molecular Formula:
C12H12Ti
InChi Key:
UGMSNGMGUIZFOA-UHFFFAOYSA-N
SMILES:
[Ti]123456789%10%11C%12C1C2C3C4C5C6%12.C71C8C9C%10C%111
1
–
1
of
1
results
1
(eta7-Cycloheptatrienyl)(eta5-cyclopentadienyl)titanium(II) 95.0+%, TCI America™
CAS: 51203-49-7 Molecular Formula: C12H12Ti Molecular Weight (g/mol): 204.10 MDL Number: MFCD04973057 InChI Key: UGMSNGMGUIZFOA-UHFFFAOYSA-N Synonym: Troticene