Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(eta7-Cycloheptatrienyl)(eta5-cyclopentadienyl)titanium(II) 95.0+%, TCI America™
SDP

Catalog No. C2991100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2991100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C2991100MG Supplier TCI America Supplier No. C2991100MG

Chemical Identifiers

CAS 51203-49-7
Molecular Formula C12H12Ti
Molecular Weight (g/mol) 204.10
MDL Number MFCD04973057
InChI Key UGMSNGMGUIZFOA-UHFFFAOYSA-N
Synonym Troticene

Specifications

Color Blue
Quantity 100 mg
UN Number 1325
Formula Weight 204.10
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (eta7-Cycloheptatrienyl)(eta5-cyclopentadienyl)titanium(II)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.