accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNH

CAS RN 51207-66-0

IUPAC Name: 1-[(pyrrolidin-2-yl)methyl]pyrrolidine
Synonyms: 1-(pyrrolidin-2-ylmethyl)pyrrolidine
Molecular Weight (g/mol): 154.26
Molecular Formula: C9H18N2
InChi Key: YLBWRMSQRFEIEB-UHFFFAOYNA-N
SMILES: C(C1CCCN1)N1CCCC1
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)