accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CNCH3OH3COCl

CAS RN 51218-45-2

IUPAC Name: 2-chloro-N-(2-ethyl-6-methylphenyl)-N-(1-methoxypropan-2-yl)acetamide
Synonyms: pennant ontrack 8E metolachlore epa pesticide chemical code 108801 caswell no. 188DD dual triple C15H22ClNO2 yibingjiacaoan metolachlor S-metolachlor
Molecular Weight (g/mol): 283.8
Molecular Formula: C15H22ClNO2
InChi Key: WVQBLGZPHOPPFO-UHFFFAOYNA-N
SMILES: CCC1=CC=CC(C)=C1N(C(C)COC)C(=O)CCl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results