accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 51219-90-0

IUPAC Name: {bis[di(²H₃)methylamino]phosphoryl}di(²H₃)methylamine
Synonyms: [didi(²H₃)methylaminophosphoryl]di(²H₃)methylamine
Molecular Weight (g/mol): 197.31
Molecular Formula: C6H18N3OP
InChi Key: GNOIPBMMFNIUFM-NBDUPMGQSA-N
SMILES: [2H]C([2H])([2H])N(C([2H])([2H])[2H])P(=O)(N(C([2H])([2H])[2H])C([2H])([2H])[2H])N(C([2H])([2H])[2H])C([2H])([2H])[2H]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results