Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexamethylphosphoramide-d18, 99.8% atom D, Thermo Scientific™

Catalog No. AC182662500
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC182662500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC182662500 Supplier Thermo Scientific Chemicals Supplier No. 182662500

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

for NMR
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51219-90-0
Molecular Formula C6H18N3OP
Molecular Weight (g/mol) 197.31
MDL Number MFCD00062430
InChI Key GNOIPBMMFNIUFM-NBDUPMGQSA-N
Synonym tris di 2h3 methylamido phosphorus, hexamethylphosphortriamide-d18, hexamethylphosphorictriamide-d18, hexamethylphosphoric triamide-d18, hexamethylphosphoricacid triamide-d18 hmpt-d18, dibis 2 h? methylaminophosphoryl bis 2 h? methylamine, n-bis bis trideuteriomethyl amino phosphoryl-1,1,1-trideuterio-n-trideuteriomethyl methanamine
PubChem CID 170997
IUPAC Name {bis[di(²H₃)methylamino]phosphoryl}di(²H₃)methylamine
SMILES [2H]C([2H])([2H])N(C([2H])([2H])[2H])P(=O)(N(C([2H])([2H])[2H])C([2H])([2H])[2H])N(C([2H])([2H])[2H])C([2H])([2H])[2H]

Specifications

Physical Form Liquid
Color Colorless
Quantity 250 mg
Water 0.05% max. (K.F.)
Deuterium (D) 99.75% min.
Linear Formula {(CD3)2N}3P(O)
Formula Weight 197.31
Chemical Name or Material Hexamethylphosphoramide-d18, 99.8 atom% D
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.